Merck
  • Distinct Connectivity and Functionality of Aldehyde Dehydrogenase 1a1-Positive Nigrostriatal Dopaminergic Neurons in Motor Learning.
Distinct Connectivity and Functionality of Aldehyde Dehydrogenase 1a1-Positive Nigrostriatal Dopaminergic Neurons in Motor Learning.
Cell reports (2019-08-01)
Junbing Wu, Justin Kung, Jie Dong, Lisa Chang, Chengsong Xie, Ahsan Habib, Sarah Hawes, Nannan Yang, Vivian Chen, Zhenhua Liu, Rebekah Evans, Bo Liang, Lixin Sun, Jinhui Ding, Jia Yu, Sara Saez-Atienzar, Beisha Tang, Zayd Khaliq, Da-Ting Lin, Weidong Le, Huaibin Cai
ABSTRACT
Parkinson's disease causes the most profound loss of the aldehyde dehydrogenase 1A1-positive (ALDH1A1+) nigrostriatal dopaminergic neuron (nDAN) subpopulation. The connectivity and functionality of ALDH1A1+ nDANs, however, remain poorly understood. Here, we show in rodent brains that ALDH1A1+ nDANs project predominantly to the rostral dorsal striatum, from which they also receive most monosynaptic inputs, indicating extensive reciprocal innervations with the striatal spiny projection neurons (SPNs). Functionally, genetic ablation of ALDH1A1+ nDANs causes severe impairments in motor skill learning, along with a reduction in high-speed walking. While dopamine replacement therapy accelerated walking speed, it failed to improve motor skill learning in ALDH1A1+ nDAN-ablated mice. Altogether, our study provides a comprehensive whole-brain connectivity map and reveals a key physiological function of ALDH1A1+ nDANs in motor skill acquisition, suggesting the motor learning processes require ALDH1A1+ nDANs to integrate diverse presynaptic inputs and supply dopamine with dynamic precision.
Sigma-Aldrich
R(+)-SKF-81297 hydrobromide, ≥98% (HPLC), solid
Sigma-Aldrich
Benserazide hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-ALDH1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Dopamine Transporter Antibody, NT, clone DAT-Nt, culture supernatant, clone DAT-Nt, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse