Merck
  • Deficiency of T-type Ca2+ channels Cav3.1 and Cav3.2 has no effect on angiotensin II-induced hypertension but differential effect on plasma aldosterone in mice.
Deficiency of T-type Ca2+ channels Cav3.1 and Cav3.2 has no effect on angiotensin II-induced hypertension but differential effect on plasma aldosterone in mice.
American journal of physiology. Renal physiology (2019-05-02)
Anne D Thuesen, Stine H Finsen, Louise L Rasmussen, Ditte C Andersen, Boye L Jensen, Pernille B L Hansen
ABSTRACT
T-type Ca2+ channel Cav3.1 promotes microvessel contraction ex vivo. It was hypothesized that in vivo, functional deletion of Cav3.1, but not Cav3.2, protects mice against angiotensin II (ANG II)-induced hypertension. Mean arterial blood pressure (MAP) and heart rate were measured continuously with chronically indwelling catheters during infusion of ANG II (30 ng·kg-1·min-1, 7 days) in wild-type (WT), Cav3.1-/-, and Cav3.2-/- mice. Plasma aldosterone and renin concentrations were measured by radioimmunoassays. In a separate series, WT mice were infused with ANG II (100 ng·kg-1·min-1) with and without the mineralocorticoid receptor blocker canrenoate. Cav3.1-/- and Cav3.2-/- mice exhibited no baseline difference in MAP compared with WT mice, but day-night variation was blunted in both Cav3.1 and Cav3.2-/- mice. ANG II increased significantly MAP in WT, Cav3.1-/-, and Cav3.2-/- mice with no differences between genotypes. Heart rate was significantly lower in Cav3.1-/- and Cav3.2-/- mice compared with control mice. After ANG II infusion, plasma aldosterone concentration was significantly lower in Cav3.1-/- compared with Cav3.2-/- mice. In response to ANG II, fibrosis was observed in heart sections from both WT and Cav3.1-/- mice and while cardiac atrial natriuretic peptide mRNA was similar, the brain natriuretic peptide mRNA increase was mitigated in Cav3.1-/- mice ANG II at 100 ng/kg yielded elevated pressure and an increased heart weight-to-body weight ratio in WT mice. Cardiac hypertrophy, but not hypertension, was prevented by the mineralocorticoid receptor blocker canrenoate. In conclusion, T-type channels Cav3.1and Cav3.2 do not contribute to baseline blood pressure levels and ANG II-induced hypertension. Cav3.1, but not Cav3.2, contributes to aldosterone secretion. Aldosterone promotes cardiac hypertrophy during hypertension.