Merck
  • Targeted Disruption of HLA Genes via CRISPR-Cas9 Generates iPSCs with Enhanced Immune Compatibility.
Targeted Disruption of HLA Genes via CRISPR-Cas9 Generates iPSCs with Enhanced Immune Compatibility.
Cell stem cell (2019-03-12)
Huaigeng Xu, Bo Wang, Miyuki Ono, Akihiro Kagita, Kaho Fujii, Noriko Sasakawa, Tatsuki Ueda, Peter Gee, Misato Nishikawa, Masaki Nomura, Fumiyo Kitaoka, Tomoko Takahashi, Keisuke Okita, Yoshinori Yoshida, Shin Kaneko, Akitsu Hotta
ABSTRACT
Induced pluripotent stem cells (iPSCs) have strong potential in regenerative medicine applications; however, immune rejection caused by HLA mismatching is a concern. B2M gene knockout and HLA-homozygous iPSC stocks can address this issue, but the former approach may induce NK cell activity and fail to present antigens, and it is challenging to recruit rare donors for the latter method. Here, we show two genome-editing strategies for making immunocompatible donor iPSCs. First, we generated HLA pseudo-homozygous iPSCs with allele-specific editing of HLA heterozygous iPSCs. Second, we generated HLA-C-retained iPSCs by disrupting both HLA-A and -B alleles to suppress the NK cell response while maintaining antigen presentation. HLA-C-retained iPSCs could evade T cells and NK cells in vitro and in vivo. We estimated that 12 lines of HLA-C-retained iPSCs combined with HLA-class II knockout are immunologically compatible with >90% of the world's population, greatly facilitating iPSC-based regenerative medicine applications.
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Pricing and availability is not currently available.