Merck
  • Validation of the CULTEX® Radial Flow System for the assessment of the acute inhalation toxicity of airborne particles.
Validation of the CULTEX® Radial Flow System for the assessment of the acute inhalation toxicity of airborne particles.
Toxicology in vitro : an international journal published in association with BIBRA (2019-03-21)
Amelie Tsoutsoulopoulos, Katrin Gohlsch, Niklas Möhle, Andreas Breit, Sebastian Hoffmann, Olaf Krischenowski, Harald Mückter, Thomas Gudermann, Horst Thiermann, Michaela Aufderheide, Dirk Steinritz
ABSTRACT
The CULTEX® Radial Flow System (RFS) is a modular in vitro system for the homogenous exposure of cells to airborne particles at the air-liquid interface (ALI). A former pre-validation study successfully demonstrated the general applicability of the CULTEX® RFS and its transferability, stability and reproducibility. Based on these results, the methodology was optimized, validated and prediction models for acute inhalation hazards were established. Cell viability of A549 cells after ALI exposure to 20 pre-selected test substances was assessed in three independent laboratories. Cytotoxicity of test substances was compared to the respective incubator controls and used as an indicator of toxicity. Substances were considered to exert an acute inhalation hazard when viability decreased below 50% (prediction model (PM) 50%) or 75% (PM 75%) at any of three exposure doses (25, 50 or 100 μg/cm2). Results were then compared to existing in vivo data and revealed an overall concordance of 85%, with a specificity of 83% and a sensitivity of 88%. Depending on the applied PM, the within-laboratory and between-laboratory reproducibility ranged from 90 to 100%. In summary, the CULTEX® RFS was proven as a transferable, reproducible and well predictive screening method for the qualitative assessment of the acute pulmonary cytotoxicity of airborne particles.