Merck
  • MYC competes with MiT/TFE in regulating lysosomal biogenesis and autophagy through an epigenetic rheostat.
MYC competes with MiT/TFE in regulating lysosomal biogenesis and autophagy through an epigenetic rheostat.
Nature communications (2019-08-11)
Ida Annunziata, Diantha van de Vlekkert, Elmar Wolf, David Finkelstein, Geoffrey Neale, Eda Machado, Rosario Mosca, Yvan Campos, Heather Tillman, Martine F Roussel, Jason Andrew Weesner, Leigh Ellen Fremuth, Xiaohui Qiu, Min-Joon Han, Gerard C Grosveld, Alessandra d'Azzo
ABSTRACT
Coordinated regulation of the lysosomal and autophagic systems ensures basal catabolism and normal cell physiology, and failure of either system causes disease. Here we describe an epigenetic rheostat orchestrated by c-MYC and histone deacetylases that inhibits lysosomal and autophagic biogenesis by concomitantly repressing the expression of the transcription factors MiT/TFE and FOXH1, and that of lysosomal and autophagy genes. Inhibition of histone deacetylases abates c-MYC binding to the promoters of lysosomal and autophagy genes, granting promoter occupancy to the MiT/TFE members, TFEB and TFE3, and/or the autophagy regulator FOXH1. In pluripotent stem cells and cancer, suppression of lysosomal and autophagic function is directly downstream of c-MYC overexpression and may represent a hallmark of malignant transformation. We propose that, by determining the fate of these catabolic systems, this hierarchical switch regulates the adaptive response of cells to pathological and physiological cues that could be exploited therapeutically.
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-4 Antibody, clone MC-813-70, clone MC-813-70, Chemicon®, from mouse
Sigma-Aldrich
Anti-Stage-Specific Embryonic Antigen-3 Antibody, clone MC-631, clone MC-631, from rat