Merck
  • Feedback inhibition of cAMP effector signaling by a chaperone-assisted ubiquitin system.
Feedback inhibition of cAMP effector signaling by a chaperone-assisted ubiquitin system.
Nature communications (2019-06-14)
Laura Rinaldi, Rossella Delle Donne, Bruno Catalanotti, Omar Torres-Quesada, Florian Enzler, Federica Moraca, Robert Nisticò, Francesco Chiuso, Sonia Piccinin, Verena Bachmann, Herbert H Lindner, Corrado Garbi, Antonella Scorziello, Nicola Antonino Russo, Matthis Synofzik, Ulrich Stelzl, Lucio Annunziato, Eduard Stefan, Antonio Feliciello
ABSTRACT
Activation of G-protein coupled receptors elevates cAMP levels promoting dissociation of protein kinase A (PKA) holoenzymes and release of catalytic subunits (PKAc). This results in PKAc-mediated phosphorylation of compartmentalized substrates that control central aspects of cell physiology. The mechanism of PKAc activation and signaling have been largely characterized. However, the modes of PKAc inactivation by regulated proteolysis were unknown. Here, we identify a regulatory mechanism that precisely tunes PKAc stability and downstream signaling. Following agonist stimulation, the recruitment of the chaperone-bound E3 ligase CHIP promotes ubiquitylation and proteolysis of PKAc, thus attenuating cAMP signaling. Genetic inactivation of CHIP or pharmacological inhibition of HSP70 enhances PKAc signaling and sustains hippocampal long-term potentiation. Interestingly, primary fibroblasts from autosomal recessive spinocerebellar ataxia 16 (SCAR16) patients carrying germline inactivating mutations of CHIP show a dramatic dysregulation of PKA signaling. This suggests the existence of a negative feedback mechanism for restricting hormonally controlled PKA activities.
Sigma-Aldrich
Anti-phospho-CREB (Ser133) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-c-Myc Tag antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CREB Antibody, Upstate®, from rabbit