Merck
  • Parp1 facilitates alternative NHEJ, whereas Parp2 suppresses IgH/c-myc translocations during immunoglobulin class switch recombination.
Parp1 facilitates alternative NHEJ, whereas Parp2 suppresses IgH/c-myc translocations during immunoglobulin class switch recombination.
The Journal of experimental medicine (2009-04-15)
Isabelle Robert, Françoise Dantzer, Bernardo Reina-San-Martin
ABSTRACT
Immunoglobulin class switch recombination (CSR) is initiated by DNA breaks triggered by activation-induced cytidine deaminase (AID). These breaks activate DNA damage response proteins to promote appropriate repair and long-range recombination. Aberrant processing of these breaks, however, results in decreased CSR and/or increased frequency of illegitimate recombination between the immunoglobulin heavy chain locus and oncogenes like c-myc. Here, we have examined the contribution of the DNA damage sensors Parp1 and Parp2 in the resolution of AID-induced DNA breaks during CSR. We find that although Parp enzymatic activity is induced in an AID-dependent manner during CSR, neither Parp1 nor Parp2 are required for CSR. We find however, that Parp1 favors repair of switch regions through a microhomology-mediated pathway and that Parp2 actively suppresses IgH/c-myc translocations. Thus, we define Parp1 as facilitating alternative end-joining and Parp2 as a novel translocation suppressor during CSR.
Product Description
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.