Merck
  • Improved success of sparse matrix protein crystallization screening with heterogeneous nucleating agents.
Improved success of sparse matrix protein crystallization screening with heterogeneous nucleating agents.
PloS one (2007-11-01)
Anil S Thakur, Gautier Robin, Gregor Guncar, Neil F W Saunders, Janet Newman, Jennifer L Martin, Bostjan Kobe
ABSTRACT
Crystallization is a major bottleneck in the process of macromolecular structure determination by X-ray crystallography. Successful crystallization requires the formation of nuclei and their subsequent growth to crystals of suitable size. Crystal growth generally occurs spontaneously in a supersaturated solution as a result of homogenous nucleation. However, in a typical sparse matrix screening experiment, precipitant and protein concentration are not sampled extensively, and supersaturation conditions suitable for nucleation are often missed. We tested the effect of nine potential heterogenous nucleating agents on crystallization of ten test proteins in a sparse matrix screen. Several nucleating agents induced crystal formation under conditions where no crystallization occurred in the absence of the nucleating agent. Four nucleating agents: dried seaweed; horse hair; cellulose and hydroxyapatite, had a considerable overall positive effect on crystallization success. This effect was further enhanced when these nucleating agents were used in combination with each other. Our results suggest that the addition of heterogeneous nucleating agents increases the chances of crystal formation when using sparse matrix screens.
Sigma-Aldrich
CM Sephadex® C-50
Pricing and availability is not currently available.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Pricing and availability is not currently available.