Merck
  • Enantioselective C-C bond formation by rhodium-catalyzed tandem ylide formation/[2,3]-sigmatropic rearrangement between donor/acceptor carbenoids and allylic alcohols.
Enantioselective C-C bond formation by rhodium-catalyzed tandem ylide formation/[2,3]-sigmatropic rearrangement between donor/acceptor carbenoids and allylic alcohols.
Journal of the American Chemical Society (2009-12-10)
Zhanjie Li, Huw M L Davies
ABSTRACT
The rhodium-catalyzed reaction of racemic allyl alcohols with methyl phenyldiazoacetate or methyl styryldiazoacetate results in a two-step process, an initial oxonium ylide formation followed by a [2,3]-sigmatropic rearrangement. This process competes favorably with the more conventional O-H insertion chemistry as long as donor/acceptor carbenoids and highly substituted allyl alcohols are used as substrates. When the reactions are catalyzed by Rh(2)(S-DOSP)(4), tertiary alpha-hydroxycarboxylate derivatives with two adjacent quaternary centers are produced with high enantioselectivity (85-98% ee).
Sigma-Aldrich
Rh2(S-PTAD)4