Merck
  • Isavuconazole: Pharmacology, Pharmacodynamics, and Current Clinical Experience with a New Triazole Antifungal Agent.
Isavuconazole: Pharmacology, Pharmacodynamics, and Current Clinical Experience with a New Triazole Antifungal Agent.
Pharmacotherapy (2015-11-26)
Jeffrey M Rybak, Kayleigh R Marx, Andrew T Nishimoto, P David Rogers
ABSTRACT
Coinciding with the continually increasing population of immunocompromised patients worldwide, the incidence of invasive fungal infections has grown over the past 4 decades. Unfortunately, infections caused by both yeasts such as Candida and molds such as Aspergillus or Mucorales remain associated with unacceptably high morbidity and mortality. In addition, the available antifungals with proven efficacy in the treatment of these infections remain severely limited. Although previously available second-generation triazole antifungals have significantly expanded the spectrum of the triazole antifungal class, these agents are laden with shortcomings in their safety profiles as well as formulation and pharmacokinetic challenges. Isavuconazole, administered as the prodrug isavuconazonium, is the latest second-generation triazole antifungal to receive U.S. Food and Drug Administration approval. Approved for the treatment of both invasive aspergillosis and invasive mucormycosis, and currently under investigation for the treatment of candidemia and invasive candidiasis, isavuconazole may have therapeutic advantages over its predecessors. With clinically relevant antifungal potency against a broad range of yeasts, dimorphic fungi, and molds, isavuconazole has a spectrum of activity reminiscent of the polyene amphotericin B. Moreover, clinical experience thus far has revealed isavuconazole to be associated with fewer toxicities than voriconazole, even when administered without therapeutic drug monitoring. These characteristics, in an agent available in both a highly bioavailable oral and a β-cyclodextrin-free intravenous formulation, will likely make isavuconazole a welcome addition to the triazole class of antifungals.
Sigma-Aldrich
Isavuconazole, ≥98% (HPLC)