Merck
  • Tumoral BRD4 expression in lymph node-negative breast cancer: association with T-bet+ tumor-infiltrating lymphocytes and disease-free survival.
Tumoral BRD4 expression in lymph node-negative breast cancer: association with T-bet+ tumor-infiltrating lymphocytes and disease-free survival.
BMC cancer (2018-07-22)
Minji Lee, Farnoosh Tayyari, Dushanthi Pinnaduwage, Jane Bayani, John M S Bartlett, Anna Marie Mulligan, Shelley B Bull, Irene L Andrulis
ABSTRACT
We previously observed that T-bet+ tumor-infiltrating T lymphocytes (T-bet+ TILs) in primary breast tumors were associated with adverse clinicopathological features, yet favorable clinical outcome. We identified BRD4 (Bromodomain-Containing Protein 4), a member of the  Bromodomain and Extra Terminal domain (BET) family, as a gene that distinguished T-bet+/high and T-bet-/low tumors. In clinical studies, BET inhibitors have been shown to suppress inflammation in various cancers, suggesting a potential link between BRD4 and immune infiltration in cancer. Hence, we examined the BRD4 expression and clinicopathological features of breast cancer. The cohort consisted of a prospectively ascertained consecutive series of women with axillary node-negative breast cancer with long follow-up. Gene expression microarray data were used to detect mRNAs differentially expressed between T-bet+/high (n = 6) and T-bet-/low (n = 41) tumors. Tissue microarrays (TMAs) constructed from tumors of 612 women were used to quantify expression of BRD4 by immunohistochemistry, which was analyzed for its association with T-bet+ TILs, Jagged1, clinicopathological features, and disease-free survival. Microarray analysis indicated that BRD4 mRNA expression was up to 44-fold higher in T-bet+/high tumors compared to T-bet-/low tumors (p = 5.38E-05). Immunohistochemical expression of BRD4 in cancer cells was also shown to be associated with T-bet+ TILs (p = 0.0415) as well as with Jagged1 mRNA and protein expression (p = 0.0171, 0.0010 respectively). BRD4 expression correlated with larger tumor size (p = 0.0049), pre-menopausal status (p = 0.0018), and high Ki-67 proliferative index (p = 0.0009). Women with high tumoral BRD4 expression in the absence of T-bet+ TILs exhibited a significantly poorer outcome (log rank test p = 0.0165) relative to other subgroups. The association of BRD4 expression with T-bet+ TILs, and T-bet+ TIL-dependent disease-free survival suggests a potential link between BRD4-mediated tumor development and tumor immune surveillance, possibly through BRD4's regulation of Jagged1 signaling pathways. Further understanding BRD4's role in different immune contexts may help to identify an appropriate subset of breast cancer patients who may benefit from BET inhibitors without the risk of diminishing the anti-tumoral immune activity.
Sigma-Aldrich
Anti-BRD4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution