Merck
  • 2-Arachidonoylglycerol, an endogenous cannabinoid receptor agonist: identification as one of the major species of monoacylglycerols in various rat tissues, and evidence for its generation through CA2+-dependent and -independent mechanisms.
2-Arachidonoylglycerol, an endogenous cannabinoid receptor agonist: identification as one of the major species of monoacylglycerols in various rat tissues, and evidence for its generation through CA2+-dependent and -independent mechanisms.
FEBS letters (1998-07-03)
S Kondo, H Kondo, S Nakane, T Kodaka, A Tokumura, K Waku, T Sugiura
ABSTRACT
The molecular species compositions of monoacylglycerols obtained from various rat tissues were examined by reverse-phase high-performance liquid chromatography (HPLC) and gas chromatography-mass spectrometry (GC-MS) analyses. We confirmed that 2-arachidonoylglycerol, an endogenous cannabinoid receptor agonist, is one of the most abundant molecular species of monoacylglycerols in the brain. Substantial amounts of 2-arachidonoylglycerol were also found in the liver, spleen, lung and kidney, but the levels were considerably lower than that in the brain. We found that a small amount of 2-arachidonoylglycerol was generated in a brain homogenate during incubation in the absence of Ca2+. Importantly, the generation of 2-arachidonoylglycerol was markedly augmented in the presence of Ca2+, suggesting that Ca2+ plays a key role in regulation of the generation of 2-arachidonoylglycerol in this tissue.
Product Description
Avanti
2-AG, Avanti Research - A Croda Brand 870450O