Merck
  • Rapid Response Fluorescence Probe Enabled In Vivo Diagnosis and Assessing Treatment Response of Hypochlorous Acid-Mediated Rheumatoid Arthritis.
Rapid Response Fluorescence Probe Enabled In Vivo Diagnosis and Assessing Treatment Response of Hypochlorous Acid-Mediated Rheumatoid Arthritis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2018-08-22)
Huan Feng, Zhiqiang Zhang, Qingtao Meng, Hongmin Jia, Yue Wang, Run Zhang
ABSTRACT
Diagnosis and early assessment of the treatment response of rheumatoid arthritis (RA) necessitates a reliable bioanalytical method for rapid, sensitive, and specific detection of the hypochlorous acid (HOCl) biomarker in inflammatory diseases. Herein, two fluorescence probes, Probe-1 and Probe-2 are developed for quantitative monitoring and visualization of inflammatory response-related HOCl levels in vitro and in vivo. In the presence of HOCl, fluorescence "OFF-ON" response is obtained for both the probes as a result of specific HOCl-triggered C=N bond cleavage reaction. Probe-1 and Probe-2 feature rapid response (<4 s), a high degree of sensitivity and selectivity toward HOCl, which allow them to be used for quantification of HOCl in a simulated physiological condition. Using Probe-2 as the probe, fluorescence imaging and flow cytometry analysis of HOCl levels in lysosome of inflammatory mimic cells, visualization of HOCl generation in endotoxin-induced inflammation of adult zebrafish and RA of mice are possible. Probe-2 exhibits high effectiveness for early assessment of the treatment response of HOCl-mediated RA in mice with an antiarthritic drug, methotrexate (MTX). The results demonstrate that Probe-2 is a powerful tool for future studies on diagnosis and monitoring treatment efficiency in a broad range of inflammatory diseases, including RA.
Sigma-Aldrich
Diaminomaleonitrile, 98%