Merck
  • Development of a Novel Q-body Using an In Vivo Site-Specific Unnatural Amino Acid Incorporation System.
Development of a Novel Q-body Using an In Vivo Site-Specific Unnatural Amino Acid Incorporation System.
Sensors (Basel, Switzerland) (2018-08-04)
Yoichi Kurumida, Nobuhiro Hayashi
ABSTRACT
A Q-body capable of detecting target molecules in solutions could serve as a simple molecular detection tool. The position of the fluorescent dye in a Q-body affects sensitivity and therefore must be optimized. This report describes the development of Nef Q-bodies that recognize Nef protein, one of the human immunodeficiency virus (HIV)'s gene products, in which fluorescent dye molecules were placed at various positions using an in vivo unnatural amino acid incorporation system. A maximum change in fluorescence intensity of 2-fold was observed after optimization of the dye position. During the process, some tryptophan residues of the antibody were found to quench the fluorescence. Moreover, analysis of the epitope indicated that some amino acid residues of the antigen located near the epitope affected the fluorescence intensity.
Sigma-Aldrich
DBCO-Cy3, for Copper-free Click Chemistry