Merck
  • Chromosomal reassignment: YACs containing both YES1 and thymidylate synthase map to the short arm of chromosome 18.
Chromosomal reassignment: YACs containing both YES1 and thymidylate synthase map to the short arm of chromosome 18.
Genomics (1993-02-01)
G A Silverman, W L Kuo, P Taillon-Miller, J W Gray
ABSTRACT
The YES1 proto-oncogene was mapped previously to human chromosome band 18q21.3 by using isotopic in situ hybridization. Using yeast artificial chromosomes (YACs) as probes and fluorescence in situ hybridization, a strong signal was detected in the region corresponding to 18p11.3. Restriction digests confirmed that the YACs contained the YES1 gene and not other cross-hybridizing, protein-tyrosine kinases. In addition, these YACs were found to contain another 18p11.32 gene, thymidylate synthase. These genes were less than 50 kb apart. Collectively, these data suggest that YES1 maps to 18p11.32 rather than to 18q21.3.
Sigma-Aldrich
YES1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution