Merck
Pulmonary pericytes regulate lung morphogenesis.
Nature communications (2018-06-24)
Katsuhiro Kato, Rodrigo Diéguez-Hurtado, Do Young Park, Seon Pyo Hong, Sakiko Kato-Azuma, Susanne Adams, Martin Stehling, Britta Trappmann, Jeffrey L Wrana, Gou Young Koh, Ralf H Adams
ABSTRACT
Blood vessels are essential for blood circulation but also control organ growth, homeostasis, and regeneration, which has been attributed to the release of paracrine signals by endothelial cells. Endothelial tubules are associated with specialised mesenchymal cells, termed pericytes, which help to maintain vessel wall integrity. Here we identify pericytes as regulators of epithelial and endothelial morphogenesis in postnatal lung. Mice lacking expression of the Hippo pathway components YAP and TAZ in pericytes show defective alveologenesis. Mutant pericytes are present in normal numbers but display strongly reduced expression of hepatocyte growth factor leading to impaired activation of the c-Met receptor, which is expressed by alveolar epithelial cells. YAP and TAZ are also required for expression of angiopoietin-1 by pulmonary pericytes, which also controls hepatocyte growth factor expression and thereby alveologenesis in an autocrine fashion. These findings establish that pericytes have important, organ-specific signalling properties and coordinate the behavior of epithelial and vascular cells during lung morphogenesis.
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Castor Oil, Ethoxylated, pH range 6.0-8.0
Sigma-Aldrich
Anti-Aquaporin 5 Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Red Blood Cell Lysing Buffer Hybri-Max, liquid, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Agarose, low gelling temperature
Sigma-Aldrich
Anti-WWTR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution