Merck
  • Orange-spotted grouper (Epinephelus coioides) toll-like receptor 22: molecular characterization, expression pattern and pertinent signaling pathways.
Orange-spotted grouper (Epinephelus coioides) toll-like receptor 22: molecular characterization, expression pattern and pertinent signaling pathways.
Fish & shellfish immunology (2012-06-12)
Xu Ding, Dan-qi Lu, Qing-hua Hou, Shui-sheng Li, Xiao-chun Liu, Yong Zhang, Hao-ran Lin
ABSTRACT
The toll-like receptors (TLRs) are an important gene family in host innate immunologic surveillance. The TLR22 gene is an essential member of the TLRs that is only found in aquatic animals and has been detected in some bony fish. Here, a TLR22 homolog, EcTLR22, was characterized in the orange-spotted grouper (Epinephelus coioides) via homology cloning. The 3321 bp full-length cDNA sequence of EcTLR22 was obtained, which included an open reading frame of 2880 bp encoding a putative peptide of 960 amino acids containing three highly typical domains with the characteristics of TLR family members. The deduced amino acid sequence of EcTLR22 showed a relatively high similarity to flounder TLR22. Phylogenetic analysis showed that the orange-spotted grouper TLR22 sequence was clustered with those of Perciforme, such as flounder and croaker. Real-time quantitative PCR analysis revealed broad expression of EcTLR22, with relatively high expression detected in the head kidney, trunk kidney, spleen, peripheral blood leukocytes (PBLs) and heart of orange-spotted grouper. After injection with Vibrio alginolyticus, there was significant up-regulation of the expression of EcTLR22 in the spleen. In evaluating unstimulated/stimulated head kidney leukocytes and spleen leukocytes, a significant increase in EcTLR22 mRNA expression was detected, which implied a sensitive immune response. Furthermore, four important molecules for signal transduction, MyD88, TRIF, TNF-α and IRF3, were chosen to analyze the role of the EcTLR22 signaling pathway in anti-pathogen responses. Upon LPS or Poly I:C challenge, expression of the four genes was induced, with an increasing tendency detected in head kidney leukocytes, suggesting that the four genes might work with EcTLR22 in host defense against pathogenic microbes.
Sigma-Aldrich
SCGF-alpha human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
SCGF-beta human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture