Merck
  • A newly identified member of the tumor necrosis factor receptor superfamily with a wide tissue distribution and involvement in lymphocyte activation.
A newly identified member of the tumor necrosis factor receptor superfamily with a wide tissue distribution and involvement in lymphocyte activation.
The Journal of biological chemistry (1997-05-30)
B S Kwon, K B Tan, J Ni, K O Oh, Z H Lee, K K Kim, Y J Kim, S Wang, R Gentz, G L Yu, J Harrop, S D Lyn, C Silverman, T G Porter, A Truneh, P R Young
ABSTRACT
The tumor necrosis factor receptor (TNFR) superfamily consists of approximately 10 characterized members of human proteins. We have identified a new member of the TNFR superfamily, TR2, from a search of an expressed sequence tag data base. cDNA cloning and Northern blot hybridization demonstrated multiple mRNA species, of which a 1.7-kilobase form was most abundant. However, TR2 is encoded by a single gene which, maps to chromosome 1p36.22-36.3, in the same region as several other members of the TNFR superfamily. The most abundant TR2 open reading frame encodes a 283-amino acid single transmembrane protein with a 36-residue signal sequence, two perfect and two imperfect TNFR-like cysteine-rich domains, and a short cytoplasmic tail with some similarity to 4-1BB and CD40. TR2 mRNA is expressed in multiple human tissues and cell lines and shows a constitutive and relatively high expression in peripheral blood T cells, B cells, and monocytes. A TR2-Fc fusion protein inhibited a mixed lymphocyte reaction-mediated proliferation suggesting that the receptor and/or its ligand play a role in T cell stimulation.
Sigma-Aldrich
HVEM-Fc human, recombinant, expressed in Hi-5 Insect cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
CD270 human, recombinant, expressed in E. coli, 0.5 mg protein/mL