Merck
  • A Dual Alkylated Peptide-ligand Enhances Affinity to Human Serum Albumin.
A Dual Alkylated Peptide-ligand Enhances Affinity to Human Serum Albumin.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2018-04-13)
Elnaz Nakhaei, Ko Takehara, Hikari Sato, Khadijah Zai, Akihiro Kishimura, Takeshi Mori, Yoshiki Katayama
ABSTRACT
Therapeutic peptides and diagnostic agents with their molecular size below the renal clearance threshold suffer from short blood circulation time. Here, we report a novel design of peptide-based ligand with a strong binding affinity to human serum albumin (HSA), which can be used as a tag to extend the blood circulation of small-size molecules. We designed ligands with dual alkyl groups connected with a negatively charged spacer. The ligands showed both higher binding affinity to HSA and a higher retention in mice blood than that of a single alkylated peptide.
Sigma-Aldrich
Tetradecylamine, 95%