Merck
  • Palmitate Activates CCL4 Expression in Human Monocytic Cells via TLR4/MyD88 Dependent Activation of NF-κB/MAPK/ PI3K Signaling Systems.
Palmitate Activates CCL4 Expression in Human Monocytic Cells via TLR4/MyD88 Dependent Activation of NF-κB/MAPK/ PI3K Signaling Systems.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-04-19)
Shihab Kochumon, Ajit Wilson, Betty Chandy, Steve Shenouda, Jaakko Tuomilehto, Sardar Sindhu, Rasheed Ahmad
ABSTRACT
Obesity is associated with adipose tissue inflammation which plays a key role in the development of insulin resistance and type 2 diabetes (T2D). Saturated free fatty acids (SFAs) levels are found to be elevated in obesity and T2D. Chemokines are known to have potent inflammatory functions in a wide range of biological processes linked to immunological disorders. Since CCL4 (Chemokine (C-C motif) ligand 4), also known as macrophage inflammatory protein-1β (MIP-1β), plays an important role in the migration of monocytes into the adipose tissue, we investigated the expression of CCL4 in monocytic cells/macrophages following activation with free fatty acid palmitate. Human monocytic cell line THP-1 and macrophages derived from THP-1 and primary monocytes were stimulated with palmitate and LPS (positive control). CCL4 expression and secretion were measured with real time RT-PCR and ELISA respectively. Signaling pathways were identified by using THP-1-XBlueTM cells, THP-1-XBlueTM-defMyD cells, anti-TLR4 mAb and TLR4 siRNA. Palmitate induces CCL4 expression at both mRNA and protein levels in human monocytic cells. Palmitate-induced CCL4 production was markedly suppressed by neutralizing anti-TLR-4 antibody. Additionally, silencing of TLR4 by siRNA also significantly suppressed the palmitate-induced up-regulation of CCL4. MyD88-deficient cells did not express CCL4 in response to palmitate treatment. Inhibition of NF-kB and MAPK pathways suppressed the palmitate mediated induction of CCL4. Moreover, induction of CCL4 was blocked by PI3 Kinase inhibitors LY294002 and wortmannin. Collectively, our results show that palmitate induces CCL4 expression via activation of the TLR4-MyD88/NF-kB/MAPK/ PI3K signaling cascade. Thus, our findings suggest that the palmitate-induced CCL4 production might be an underlying mechanism of metabolic inflammation.
Sigma-Aldrich
6-Carboxyfluorescein, ≥96% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR4