Merck
  • Chemistry-First Approach for Nomination of Personalized Treatment in Lung Cancer.
Chemistry-First Approach for Nomination of Personalized Treatment in Lung Cancer.
Cell (2018-04-24)
Elizabeth A McMillan, Myung-Jeom Ryu, Caroline H Diep, Saurabh Mendiratta, Jean R Clemenceau, Rachel M Vaden, Ju-Hwa Kim, Takashi Motoyaji, Kyle R Covington, Michael Peyton, Kenneth Huffman, Xiaofeng Wu, Luc Girard, Yeojin Sung, Pei-Hsaun Chen, Prema L Mallipeddi, Joo Young Lee, Jordan Hanson, Sukesh Voruganti, Yunku Yu, Sunho Park, Jessica Sudderth, Christopher DeSevo, Donna M Muzny, HarshaVardhan Doddapaneni, Adi Gazdar, Richard A Gibbs, Tae-Hyun Hwang, John V Heymach, Ignacio Wistuba, Kevin R Coombes, Noelle S Williams, David A Wheeler, John B MacMillan, Ralph J Deberardinis, Michael G Roth, Bruce A Posner, John D Minna, Hyun Seok Kim, Michael A White
ABSTRACT
Diversity in the genetic lesions that cause cancer is extreme. In consequence, a pressing challenge is the development of drugs that target patient-specific disease mechanisms. To address this challenge, we employed a chemistry-first discovery paradigm for de novo identification of druggable targets linked to robust patient selection hypotheses. In particular, a 200,000 compound diversity-oriented chemical library was profiled across a heavily annotated test-bed of >100 cellular models representative of the diverse and characteristic somatic lesions for lung cancer. This approach led to the delineation of 171 chemical-genetic associations, shedding light on the targetability of mechanistic vulnerabilities corresponding to a range of oncogenotypes present in patient populations lacking effective therapy. Chemically addressable addictions to ciliogenesis in TTC21B mutants and GLUT8-dependent serine biosynthesis in KRAS/KEAP1 double mutants are prominent examples. These observations indicate a wealth of actionable opportunities within the complex molecular etiology of cancer.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
SW157765, ≥98% (HPLC)