Merck
  • Part-per-trillion level determination of antifouling pesticides and their byproducts in seawater samples by off-line solid-phase extraction followed by high-performance liquid chromatography-atmospheric pressure chemical ionization mass spectrometry.
Part-per-trillion level determination of antifouling pesticides and their byproducts in seawater samples by off-line solid-phase extraction followed by high-performance liquid chromatography-atmospheric pressure chemical ionization mass spectrometry.
Journal of chromatography. A (2000-06-28)
K Martínez, I Ferrer, D Barceló
ABSTRACT
A new method for the simultaneous determination of antifouling pesticides and some of their byproducts such as dichlofluanid, diuron and its byproducts [demethyldiuron and 1-(3,4-dichlorophenyl)urea], (2-thiocyanomethylthio)ben: zothiazole, chlorothalonil, Sea-nine 211, Irgarol 1051 and one of its byproducts (2-methylthio-4-tert.-butylamino-s-triazine) in seawater was developed. The extraction of these compounds from the filtered seawater samples was performed off-line with different solid-phase extraction sorbents using (I) a 500 mg graphitized carbon black cartridge (ENVI-Carb) and (II) 200 mg polymeric cartridges (LiChrolut EN and Isolute ENV+) and passing 500 ml of the sample through these cartridges. The detection was carried out by reversed-phase high-performance liquid chromatography coupled with atmospheric pressure chemical ionization mass spectrometry both in the negative and positive ion modes. The recovery ranged from 76 to 96% for the whole antifouling group with the ENVI-Carb cartridges and the detection limit was at the part-per-trillion level except for TCMTB. The method utilizing the polymeric cartridge proved to be very useful, time saving and with good recoveries when only Irgarol and its byproduct, Sea-nine 211 and diuron and its byproducts, have to be analyzed. The different cartridges were applied to the analysis of these pesticides in different marinas of the Catalan coast; diuron, dichlofluanid, Sea-nine 211, Irgarol as well as demethyldiuron and the Irgarol byproduct being the must ubiquitous pollutants. Maximum concentration levels were 2-3.5 microg/l of diuron and Sea-nine 211, respectively.
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS