![Verapamil Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/286/113/80213e13-879e-445d-b1a1-451274f513a7/640/80213e13-879e-445d-b1a1-451274f513a7.jpg)
Verapamil Related Compound B
![Verapamil Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/286/113/80213e13-879e-445d-b1a1-451274f513a7/640/80213e13-879e-445d-b1a1-451274f513a7.jpg)
Synonym(s):
4-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylbutyronitrile monohydrochloride
Empirical Formula (Hill Notation):
C26H36N2O4 · HCl
CAS Number:
Molecular Weight:
477.04
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2