![Valeraldehyde-DNPH solution certified reference material, 100 μg/mL in acetonitrile (as the aldehyde), ampule of 1 mL](/deepweb/assets/sigmaaldrich/product/images/104/414/928be3b1-74b6-44d1-9e44-cdf197e4b8b4/640/928be3b1-74b6-44d1-9e44-cdf197e4b8b4.jpg)
Valeraldehyde-DNPH solution
![Valeraldehyde-DNPH solution certified reference material, 100 μg/mL in acetonitrile (as the aldehyde), ampule of 1 mL](/deepweb/assets/sigmaaldrich/product/images/104/414/928be3b1-74b6-44d1-9e44-cdf197e4b8b4/640/928be3b1-74b6-44d1-9e44-cdf197e4b8b4.jpg)
Synonym(s):
Valeraldehyde-2,4-dinitrophenylhydrazone solution
CAS Number:
Products
Brand
Product Line
Format
Quality Segment
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing