![TSKgel® SuperAW3000 HPLC Column phase hydroxylated methacrylate, L × I.D. 15 cm × 6 mm, 4 μm particle size](/deepweb/assets/sigmaaldrich/product/images/419/839/0fe9f6fb-0243-4d15-a072-858c71260659/640/0fe9f6fb-0243-4d15-a072-858c71260659.jpg)
TSKgel® SuperAW3000 (4 µm) HPLC Columns
![TSKgel® SuperAW3000 HPLC Column phase hydroxylated methacrylate, L × I.D. 15 cm × 6 mm, 4 μm particle size](/deepweb/assets/sigmaaldrich/product/images/419/839/0fe9f6fb-0243-4d15-a072-858c71260659/640/0fe9f6fb-0243-4d15-a072-858c71260659.jpg)
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Column
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing