![TSKgel® GMHXL-L HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 6 μm particle size](/deepweb/assets/sigmaaldrich/product/images/363/550/1a73f1fd-4320-410b-b949-fe2a71ddccf8/640/1a73f1fd-4320-410b-b949-fe2a71ddccf8.jpg)
TSKgel® GMHXL-L (6 µm) HPLC Columns
![TSKgel® GMHXL-L HPLC Column phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 6 μm particle size](/deepweb/assets/sigmaaldrich/product/images/363/550/1a73f1fd-4320-410b-b949-fe2a71ddccf8/640/1a73f1fd-4320-410b-b949-fe2a71ddccf8.jpg)
Synonym(s):
TSKgel® G1000HXL HPLC Column for Light Scattering Applications
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing