Skip to Content
Merck
TSKgel® CM-3SW HPLC Column phase carboxymethyl, L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size

TSKgel® CM-3SW (10 µm) HPLC Columns

TSKgel® CM-3SW HPLC Column phase carboxymethyl, L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size
Synonym(s):
TSKgel® CM-SW Columns

Products

Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1