![TSKgel® BioAssist® S HPLC Column phase sulfonic acid, L × I.D. 10 cm × 10 mm, 13 μm particle size, PEEK hardware](/deepweb/assets/sigmaaldrich/product/images/329/010/0555e4ab-18c8-462b-b236-93c648209b3a/640/0555e4ab-18c8-462b-b236-93c648209b3a.jpg)
TSKgel® BioAssist® S (13 µm) HPLC Columns
![TSKgel® BioAssist® S HPLC Column phase sulfonic acid, L × I.D. 10 cm × 10 mm, 13 μm particle size, PEEK hardware](/deepweb/assets/sigmaaldrich/product/images/329/010/0555e4ab-18c8-462b-b236-93c648209b3a/640/0555e4ab-18c8-462b-b236-93c648209b3a.jpg)
Synonym(s):
TSKgel® BioAssist® S HPLC Column
Products
Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing