Skip to Content
Merck
Trityl chloride, polymer-bound 200-400 mesh, extent of labeling: 1.0-1.8 mmol/g loading, 1 % cross-linked

Trityl chloride, polymer-bound

Trityl chloride, polymer-bound 200-400 mesh, extent of labeling: 1.0-1.8 mmol/g loading, 1 % cross-linked
Synonym(s):
Chlorotriphenylmethane, polymer-bound, Polystyrene crosslinked with divinylbenzene, α-chlorobenzhydrylated, Triphenylchloromethane, polymer-bound, Triphenylmethyl chloride

Products

Brand
Physical Form
Quality Segment
Application
Cleavage Condition
Linker
Chain Type
Reaction Type
Reactivity