Skip to Content
Merck
Triethyl 2-acetylcitrate Pharmaceutical Secondary Standard; Certified Reference Material

Triethyl 2-acetylcitrate

Triethyl 2-acetylcitrate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Acetyl triethyl citrate
Linear Formula:
CH3COOC(COOCH2CH3)(CH2COOCH2CH3)2
CAS Number:
Molecular Weight:
318.32
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2