Skip to Content
Merck
Triclopyr 2-butoxyethylester PESTANAL®, analytical standard

Triclopyr 2-butoxyethylester

Triclopyr 2-butoxyethylester PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C13H16Cl3NO4
CAS Number:
Molecular Weight:
356.63
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing