
Thiothixene

Synonym(s):
cis-Thiothixene, N,N-Dimethyl-9-[3-(4-methyl-1-piperazinyl)propylidene]thioxanthene-2-sulfonamide
Empirical Formula (Hill Notation):
C23H29N3O2S2
CAS Number:
Molecular Weight:
443.63
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing