
Theaspirane

Synonym(s):
(±)-Theaspirane, 2,6,10,10-Tetramethyl-1-oxaspiro[4.5]dec-6-ene
Empirical Formula (Hill Notation):
C13H22O
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing