Skip to Content
CZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Testosterone propionate
Synonym(s):
17β-Hydroxy-4-androsten-3-one 17-propionate, 17β-Propionyloxy-4-androsten-3-one, 4-Androsten-17β-ol-3-one 17-propionate
Empirical Formula (Hill Notation):
C
22
H
32
O
3
CAS Number:
57-85-2
Molecular Weight:
344.49
EC Number:
200-351-1
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
86541
Testosterone propionate,
tested according to Ph. Eur.
Expand
View Pricing
T0300000
Testosterone propionate,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
BP702
Testosterone propionate,
British Pharmacopoeia (BP) Reference Standard
Expand
View Pricing