Skip to Content
Merck
Testosterone enanthate ≥98% (HPLC)

Testosterone enanthate

Testosterone enanthate ≥98% (HPLC)
Synonym(s):
(17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, 4-Androstene-17β-ol-3-one enanthate, Heptanoic acid ester with testosterone, Testosterone heptanoate
Empirical Formula (Hill Notation):
C26H40O3
CAS Number:
Molecular Weight:
400.59

Products

Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing