
Testosterone acetate

Synonym(s):
17β-Acetoxy-4-androsten-3-one, 17β-Hydroxy-4-androsten-3-one 17-acetate, 4-Androsten-17β-ol-3-one 17-acetate, Deposteron
Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
Products
Brand
Formula Weight
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing