Skip to Content
Merck
vibrant-m

Terbutaline impurity C

vibrant-m
Synonym(s):
1-(3,5-Dihydroxyphenyl)-2-[(1,1-dimethylethyl)amino]ethanone, Terbutalone
Empirical Formula (Hill Notation):
C12H17NO3
CAS Number:
Molecular Weight:
223.27

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade