Teniposide
Synonym(s):
4′-Dimethyl-9-(4,6-O-2-thenyid)-epipodophyllotoxin, Tenoposide, VM-26
Empirical Formula (Hill Notation):
C32H32O13S
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing