Skip to Content
Merck
Tamoxifen citrate salt ≥99%

Tamoxifen citrate salt

Tamoxifen citrate salt ≥99%
Synonym(s):
(Z)-1-(p-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene
Empirical Formula (Hill Notation):
C26H29NO · C6H8O7
CAS Number:
Molecular Weight:
563.64
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing