Skip to Content
Merck
vibrant-m

Tacrolimus 8-propyl analog

vibrant-m
Synonym(s):
Tsukubamycin B
Empirical Formula (Hill Notation):
C44H71NO12
CAS Number:
Molecular Weight:
806.03

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1