![SUPELCOGEL™ Pb Guard Column 9 μm particle size, L × I.D. 5 cm × 4.6 mm](/deepweb/assets/sigmaaldrich/product/images/121/475/0f87d71f-a019-4beb-a18d-7520edd6a879/640/0f87d71f-a019-4beb-a18d-7520edd6a879.jpg)
SUPELCOGEL™ Pb (9 µm) HPLC Columns
![SUPELCOGEL™ Pb Guard Column 9 μm particle size, L × I.D. 5 cm × 4.6 mm](/deepweb/assets/sigmaaldrich/product/images/121/475/0f87d71f-a019-4beb-a18d-7520edd6a879/640/0f87d71f-a019-4beb-a18d-7520edd6a879.jpg)
Synonym(s):
SUPELCOGEL™ 6% Cross Linked HPLC Columns
Products
Product Category
Brand
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Column Material
Package Size
Particle Size (µm)
Technique
Showing 1-3 of 3