Skip to Content
Merck
Sucrose monolaurate BioXtra, ≥97.0% (TLC)

Sucrose monolaurate

Sucrose monolaurate BioXtra, ≥97.0% (TLC)
Synonym(s):
n-Dodecanoylsucrose, Lauric acid sucrose ester, Sucrose monododecanoate
Empirical Formula (Hill Notation):
C24H44O12
CAS Number:
Molecular Weight:
524.60
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing