
Sodium iodohippurate

Synonym(s):
Sodium o-iodohippurate
Empirical Formula (Hill Notation):
C9H8INO3
CAS Number:
Molecular Weight:
305.07
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing