Skip to Content
Merck
Sodium 3-(benzothiazol-2-ylthio)-1-propanesulfonate technical, ≥90% (NT)

Sodium 3-(benzothiazol-2-ylthio)-1-propanesulfonate

Sodium 3-(benzothiazol-2-ylthio)-1-propanesulfonate technical, ≥90% (NT)
Synonym(s):
3-(Benzothiazol-2-ylthio)-1-propanesulfonic acid sodium salt, ZPS
Empirical Formula (Hill Notation):
C10H10NNaO3S3
CAS Number:
Molecular Weight:
311.38
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing