
Sennidin B

Synonym(s):
(9S)-9-[(9R)-2-carboxy-4,5-dihydroxy-10-oxo-9H-anthracen-9-yl]-4,5-dihydroxy-10-oxo-9H-anthracene-2-carboxylic acid
Empirical Formula (Hill Notation):
C30H18O10
CAS Number:
Molecular Weight:
538.46
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing