Skip to Content
Merck
Riboflavin 5′-monophosphate sodium salt hydrate ≥95% (HPLC)

Riboflavin 5′-monophosphate sodium salt hydrate

Riboflavin 5′-monophosphate sodium salt hydrate ≥95% (HPLC)
Empirical Formula (Hill Notation):
C17H20N4O9P · xNa+ · yH2O
Molecular Weight:
455.34 (anhydrous free acid basis)
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing