
Rhaponticin

Synonym(s):
Rhapontin, 3,3′,5-Trihydroxy-4′-methoxystilbene 3-O-β-D-glucoside, NSC 43321, Rhaponticin
Empirical Formula (Hill Notation):
C21H24O9
CAS Number:
Molecular Weight:
420.41
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing