Skip to Content
Merck
(R)-(−)-2-(2,5-Dihydrophenyl)glycine 98%

(R)-(−)-2-(2,5-Dihydrophenyl)glycine

(R)-(−)-2-(2,5-Dihydrophenyl)glycine 98%
Synonym(s):
(R)-α-Amino-1,4-cyclohexadiene-1-acetic acid
Empirical Formula (Hill Notation):
C8H11NO2
CAS Number:
Molecular Weight:
153.18
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing