![Quizalofop-P certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/390/680/53ad4bc0-f76c-4fc3-bed8-9af39b43013f/640/53ad4bc0-f76c-4fc3-bed8-9af39b43013f.png)
Quizalofop-P
![Quizalofop-P certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/390/680/53ad4bc0-f76c-4fc3-bed8-9af39b43013f/640/53ad4bc0-f76c-4fc3-bed8-9af39b43013f.png)
Synonym(s):
(R)-2-[4-(6-Chloro-2-quinoxalinyloxy)phenoxy]propionic acid
Empirical Formula (Hill Notation):
C17H13ClN2O4
CAS Number:
Molecular Weight:
344.75
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2