
Quetiapine Related Compound G

Synonym(s):
Dibenzo[b,f][1,4]thiazepin-11(10H)-one, NSC 653252
Empirical Formula (Hill Notation):
C13H9NOS
CAS Number:
Molecular Weight:
227.28
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2